Skip to Content
Merck
All Photos(1)

Key Documents

B8776

Sigma-Aldrich

Botulinum Toxin A from Clostridium botulinum

lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

form

lyophilized powder

shipped in

dry ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibits release of catecholamines from adrenal medulla, acetylcholine at neuromuscular junctions.
Neurotoxin that inhibits release of catecholamines from adrenal medulla; blocks release of acetylcholine at neuromuscular junctions.

Physical form

Lyophilized powder containing 20 mM HEPES and 1.25% lactose when reconstituted with the volume of water or buffer indicated on the Certifcate of Analysis.

Preparation Note

When toxin is used for in vitro studies, it is preincubated in a buffer containing 5 mM dithiothreitol for 30 minutes in order to reduce and thereby activate the toxin.

Reconstitution

Reconstitute each 100 μg vial with 100 μL of appropriate buffer. Upon reconstitution, the vial will contain 20 mM HEPES and 1.25% lactose. To insure full recovery of toxin from the vial, include 1 mg/mL BSA in the reconstitution buffer.

Other Notes

Handle the toxin gently. Do not vortex. Use sufficient volume of buffer to wash down the walls and stopper. Read all handling information found on SDS prior to reconstitution.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 1 Oral

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E López-Alonso et al.
Neuroscience letters, 196(1-2), 37-40 (1995-08-18)
Giant liposomes were made from a mixture of asolectin phospholipid vesicles and presynaptic plasma membranes isolated from Torpedo cholinergic nerve endings. Acetylcholine filled giant liposomes were able to release neurotransmitter upon stimulation by the Ca2+ ionophore A23187 and Ca2+. Botulinum

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service