Skip to Content
Merck
All Photos(1)

Documents

B2050

Sigma-Aldrich

3-Bromo-7-nitroindazole

≥98% (TLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4BrN3O2
CAS Number:
Molecular Weight:
242.03
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥98% (TLC)

form

powder

color

light yellow

solubility

ethanol: 25 mg/mL
DMSO: soluble

storage temp.

2-8°C

SMILES string

[O-][N+](=O)c1cccc2c(Br)n[nH]c12

InChI

1S/C7H4BrN3O2/c8-7-4-2-1-3-5(11(12)13)6(4)9-10-7/h1-3H,(H,9,10)

InChI key

NFSTZPMYAZRZPC-UHFFFAOYSA-N

Biochem/physiol Actions

A potent inhibitor of all nitric oxide synthase isoforms. Shows substantial neuroprotective effects against hypoxic damage (such as stroke) and animal models of Parkinson′s disease. Crystallographic studies show that inhibition of eNOS is due to an induced cascade of conformation changes that ultimately dissociates the tetrahydrobiopterin cofactor from the enzyme.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saurav Bhowmick et al.
Free radical biology & medicine, 113, 203-211 (2017-10-01)
Cerebral ischemia-reperfusion (I/R) injury initiates a cascade of events, generating nitric oxide (NO) and superoxide(O
K Wada et al.
Journal of neurosurgery, 89(5), 807-818 (1998-11-17)
Although nitric oxide (NO) has been shown to play an important role in the pathophysiological process of cerebral ischemia, its contribution to the pathogenesis of traumatic brain injury (TBI) remains to be clarified. The authors investigated alterations in constitutive nitric
N E Stagliano et al.
Brain research, 759(1), 32-40 (1997-06-06)
Although nitric oxide (NO) has been shown to play an important role in the pathophysiology of cerebral ischemia, its contribution to the pathogenesis of experimentally induced thromboembolic stroke is unknown. In this study, we pharmacologically manipulated NO levels in the
M J O'Neill et al.
Brain research, 760(1-2), 170-178 (1997-06-20)
Recent studies have shown that the novel antioxidant LY231617 protects against ischaemia-induced neuronal damage in rat models of global cerebral ischaemia. In the present studies we have examined the effects of LY231617 in the gerbil model of global cerebral ischaemia.
P A Bland-Ward et al.
Life sciences, 57(11), PL131-PL135 (1995-01-01)
The effect of 7-nitro indazole (7-NI) and a range of substituted indazole derivatives on nitric oxide synthase (NOS) enzyme activity in homogenates of rat cerebellum, bovine endothelial cells and lung from endotoxin-pretreated rats was investigated. 3-bromo 7-nitro indazole was either

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service