Skip to Content
Merck
All Photos(1)

Key Documents

A3317

Sigma-Aldrich

Arg-Thr-Lys-Arg-Ser-Gly-Ser-Val-Tyr-Glu-Pro-Leu-Lys-Ile

≥97% (HPLC), powder

Synonym(s):

Malantide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C72H124N22O21
CAS Number:
Molecular Weight:
1633.89
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

Assay

≥97% (HPLC)

form

powder

composition

Peptide content, ≥65%

color

white

solubility

H2O: 1 mg/mL

UniProt accession no.

storage temp.

−20°C

SMILES string

CCC(C)C(NC(=O)C(CCCCN)NC(=O)C(CC(C)C)NC(=O)C1CCCN1C(=O)C(CCC(O)=O)NC(=O)C(Cc2ccc(O)cc2)NC(=O)C(NC(=O)C(CO)NC(=O)CNC(=O)C(CO)NC(=O)C(CCCNC(N)=N)NC(=O)C(CCCCN)NC(=O)C(NC(=O)C(N)CCCNC(N)=N)C(C)O)C(C)C)C(O)=O

Gene Information

human ... PHKB(5257)

Substrates

High-affinity substrate for cAMP-dependent protein kinase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K J Murray et al.
The Biochemical journal, 267(3), 703-708 (1990-05-01)
1. The cyclic AMP-dependent protein kinase activity-ratio assay was investigated by comparing histone and a synthetic peptide, malantide [Malencik & Anderson (1983) Anal. Biochem. 132, 32-40], as substrates. 2. In several tissues the activity ratio was higher when assayed with
Cyclic AMP-dependent protein kinase activity ratio assay.
K J Murray
Methods in molecular biology (Clifton, N.J.), 41, 113-122 (1995-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service