Skip to Content
Merck
All Photos(1)

Key Documents

L1013

Sigma-Aldrich

Lowry Reagent

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352200

IVD

for in vitro diagnostic use

Other Notes

Dry powder mixture containing Lowry reagent with sodium dodecyl sulfate.

Reconstitution

Reconstitute to 40 mL with deionized water.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Customers Also Viewed

    Slide 1 of 1

    1 of 1

    Protein measurement with the Folin phenol reagent.
    O H LOWRY et al.
    The Journal of biological chemistry, 193(1), 265-275 (1951-11-01)

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service