Skip to Content
Merck
All Photos(4)

Key Documents

477R-1

Sigma-Aldrich

MAGEC2 (EP405) Rabbit Monoclonal Primary Antibody

Synonym(s):

Melanoma-associated antigen C2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

100
500

conjugate

unconjugated

antibody form

culture supernatant

antibody product type

primary antibodies

clone

EP405, monoclonal

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL concentrate (477R-14)
vial of 0.1 mL concentrate Research Use Only (477R-14-RUO)
vial of 0.5 mL concentrate (477R-15)
vial of 1.0 mL concentrate (477R-16)
vial of 1.0 mL concentrate Research Use Only (477R-16-RUO)
vial of 1.0 mL pre-dilute Research Use Only (477R-17-RUO)
vial of 1.0 mL pre-dilute ready-to-use (477R-17)
vial of 7.0 mL pre-dilute ready-to-use (477R-18)
vial of 7.0 mL pre-dilute ready-to-use Research Use Only (477R-18-RUO)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:25-1:50 (concentrated)

isotype

IgG

control

seminoma

shipped in

wet ice

storage temp.

2-8°C

visualization

nuclear

General description

Melanoma-associated antigen C2 (MAGEC2), encoded by the testis cancer gene CT10, belongs to a family of proteins that bind E3 RING ubiquitin ligases via a common MAGE homology domain.1-3 The MAGE-RING complexes that are formed enhance the activity of the ubiquitin ligase binding partners. In vitro studies suggest that MAGEC2, which binds to TRIM28, is involved in tumorigenesis through the increased degradation of p53.4 In germ cell tumors, aberrant expression of MAGEC2 is exhibited in seminomas, but is absent in yolk sac tumor and embryonal carcinoma.5-7

Quality

United States - IVD
Canada - RUO
European Union - IVD
Japan - RUO

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Download the latest released IFU
Note: This IFU may not apply to your specific product lot.

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A O Güre et al.
International journal of cancer, 85(5), 726-732 (2000-03-04)
Assays relying on humoral or T-cell-based recognition of tumor antigens to identify potential targets for immunotherapy have led to the discovery of a significant number of immunogenic gene products, including cancer-testis (CT) antigens predominantly expressed in cancer cells and male
Annikka Weissferdt et al.
Human pathology, 46(3), 376-383 (2015-01-13)
Primary mediastinal seminomas are unusual tumors that can present in a pure form or as part of a mixed germ cell tumor. Contrary to testicular seminomas, little is known about the expression of novel immunohistochemical markers in mediastinal seminomas. This
Ran Zhuang et al.
Cancer immunity, 6, 7-7 (2006-04-06)
CT10/MAGE-C2 is a recently identified antigen that, typically of cancer/testis (CT) antigens, can be found in various malignant tumors and in normal adult testis. As with many other CT antigens, our knowledge is based mainly on mRNA expression data. In
Peter K Bode et al.
Modern pathology : an official journal of the United States and Canadian Academy of Pathology, Inc, 24(6), 829-835 (2011-07-23)
Melanoma-associated gene C2 (MAGEC2) is a recently identified cancer testis antigen expressed in normal testicular and placental tissue. It has been detected in some human carcinomas, but its expression in primary testicular germ cell tumors is unknown. Immunohistochemistry was used
Anna K Lee et al.
Journal of molecular biology, 429(8), 1114-1142 (2017-03-17)
Melanoma antigen (MAGE) genes are conserved in all eukaryotes and encode for proteins sharing a common MAGE homology domain. Although only a single MAGE gene exists in lower eukaryotes, the MAGE family rapidly expanded in eutherians and consists of more

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service