Skip to Content
Merck
All Photos(1)

Key Documents

42566

Sigma-Aldrich

2,3-Dipalmitoyl-sn-glycero-1-phosphocholine

≥95.0% (TLC)

Synonym(s):

1-sn-Phosphatidylcholine, 2,3-dipalmitoyl, D-β,γ-Dipalmitoyl-α-lecithin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H80NO8P
CAS Number:
Molecular Weight:
734.04
Beilstein:
5787775
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95.0% (TLC)

form

powder

functional group

phospholipid

lipid type

phosphoglycerides

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@@H](COP([O-])(=O)OCC[N+](C)(C)C)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C40H80NO8P/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-39(42)46-36-38(37-48-50(44,45)47-35-34-41(3,4)5)49-40(43)33-31-29-27-25-23-21-19-17-15-13-11-9-7-2/h38H,6-37H2,1-5H3/t38-/m0/s1

InChI key

KILNVBDSWZSGLL-LHEWISCISA-N

Biochem/physiol Actions

2,3-Dipalmitoyl-sn-glycerol-1-phosphocholine is a major constituent of lung surfactant, a monolayer reducing lung surface tension and keeping the lung from collapsing when exhaling .

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Goerke
Biochimica et biophysica acta, 1408(2-3), 79-89 (1998-11-14)
This review briefly notes recent findings important for understanding the surface mechanical functions of pulmonary surfactant. Currently known surfactant-specific proteins and lipids are discussed, with an eye to their possible functions. Competing models of the alveolar subphase life cycle of
Peggy Scholtysek et al.
Soft matter, 11(30), 6106-6117 (2015-07-03)
Cholesterol (Ch) linked to a linear-hyperbranched block copolymer composed of poly(ethylene glycol) (PEG) and poly(glycerol) (hbPG) was investigated for its membrane anchoring properties. Two polyether-based linear-hyperbranched block copolymers with and without a covalently attached rhodamine fluorescence label (Rho) were employed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service