Skip to Content
Merck
All Photos(1)

Key Documents

17884

Sigma-Aldrich

3-(2-Furoyl)quinoline-2-carboxaldehyde

≥95% (HPLC), suitable for fluorescence, BioReagent

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$2,320.00

$2,320.00


Estimated to ship on20 April 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$2,320.00

About This Item

Empirical Formula (Hill Notation):
C15H9NO3
CAS Number:
Molecular Weight:
251.24
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.32

$2,320.00


Estimated to ship on20 April 2025


Request a Bulk Order

product line

BioReagent

Quality Level

Assay

≥95% (HPLC)

form

powder

solubility

methanol: soluble

fluorescence

λex 486 nm; λem ~600 nm in 0.1 M sodium borate pH 9.0 (after derivatization with glycine)

suitability

suitable for fluorescence

storage temp.

2-8°C

SMILES string

[H]C(=O)c1nc2ccccc2cc1C(=O)c3ccco3

InChI

1S/C15H9NO3/c17-9-13-11(15(18)14-6-3-7-19-14)8-10-4-1-2-5-12(10)16-13/h1-9H

InChI key

PNCHURHVMDRFTR-UHFFFAOYSA-N

General description

3-(2-Furoyl)quinoline-2-carboxaldehyde is a neutral fluorogenic probe for amines for the picomolar assay of proteins by capillary electrophoresis (CE).[1][2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cuiru Zhu et al.
Analytical chemistry, 79(2), 765-768 (2007-01-16)
In two-dimensional capillary electrophoresis, a sample undergoes separation in the first dimension capillary by sieving electrophoresis. Fractions are periodically transferred across an interface into a second dimension capillary, where components are further resolved by micellar electrokinetic capillary electrophoresis. Previous instruments
D M Pinto et al.
Electrophoresis, 16(4), 534-540 (1995-04-01)
The fluorescent labeling of peptides at concentrations as low as 10(-8) M can be achieved by using a solid-phase reactor. Using oxidized insulin chain B as a test peptide, we demonstrate the use of an Immobilon CD membrane to capture
Maria Teresa Veledo et al.
Analytica chimica acta, 658(1), 81-86 (2010-01-20)
Capillary electrophoresis (CE) coupled with laser-induced fluorescence detection (LIF) has allowed to obtain protein fingerprints, which have demonstrated to be useful in microorganisms characterization. In this work, protein fingerprints of two species of Staphylococcus grown in different culture media and
Cristina Pelaez-Lorenzo et al.
Methods in molecular biology (Clifton, N.J.), 984, 237-251 (2013-02-07)
This chapter describes a complete procedure for obtaining protein fingerprints of microorganisms using capillary electrophoresis (CE) with laser-induced fluorescence detection (LIF). Staphylococcus aureus, a human pathogen responsible of frequent and resistant infections, is used as model microorganism to show the
Roza Wojcik et al.
Journal of chromatography. A, 1194(2), 243-248 (2008-05-16)
3-(2-Furoyl)quinoline-2-carboxaldehyde (FQ), Chromeo P465, and Chromeo P503 are weakly fluorescent reagents that react with primary amines to produce fluorescent products. We studied the reaction of these reagents with alpha-lactalbumin by mass spectrometry. The reaction generated a set of products by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service