Skip to Content
Merck
All Photos(1)

Documents

24194

Sigma-Aldrich

Ethanol (denatured)

≥98% (GC), absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC)

Synonym(s):

Ethyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2OH
CAS Number:
Molecular Weight:
46.07
Beilstein:
1718733
MDL number:
UNSPSC Code:
15101802
PubChem Substance ID:
NACRES:
NA.21

product name

Ethanol, absolute, denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC), ≥98% (GC)

vapor density

1.59 (vs air)

vapor pressure

44.6 mmHg ( 20 °C)

grade

absolute

Assay

≥98% (GC)

form

liquid

autoignition temp.

683 °F

quality

denaturated with 0.5-1.5 Vol.% 2-butanone and approx. 0.001% Bitrex (GC)

shelf life

~5 yr

expl. lim.

19 %, 60 °F

availability

available only in Switzerland

impurities

≤0.002% free acid (as CH3COOH)
≤0.2% water (Karl Fischer)
0.001% bitrex
0.5-1.5 % (v/v) methyl ethyl ketone (GC)

refractive index

n20/D 1.3600 (lit.)

pH

7.0 (20 °C, 10 g/L)

bp

78 °C (lit.)

mp

−114 °C (lit.)

solubility

water: soluble (completely)

density

0.789 g/mL at 25 °C (lit.)

suitability

corresponds for identity

format

neat

SMILES string

CCO

InChI

1S/C2H6O/c1-2-3/h3H,2H2,1H3

InChI key

LFQSCWFLJHTTHZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Footnote

1Distribution and use of ethyl alcohol in Switzerland require a license from the Swiss Federal Alcohol Administration

Disclaimer

Distribution and use of ethyl alcohol require a license from the Swiss Federal Alcohol Administration

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

55.4 °F - closed cup

Flash Point(C)

13 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Physicochemical Properties of SnO2-Sb2 O 5 Films Prepared by the Spray Pyrolysis Technique.
Correa-Lozano B, et al.
Journal of the Electrochemical Society, 143(1), 203-209 (1996)
Spectrophotometric characteristics of chlorophylls a and b and their pheophytins in ethanol.
J F Wintermans et al.
Biochimica et biophysica acta, 109(2), 448-453 (1965-11-29)
50-Hz extremely low frequency electromagnetic fields enhance cell proliferation and DNA damage: possible involvement of a redox mechanism.
Wolf FI, et al.
Biochimica et Biophysica Acta - Molecular Cell Research, 1743(1), 120-129 (2005)
Synthesis of ZnO nanorod and the annealing effect on its photoluminescence property.
Wu L, et al.
Opt. Mater., 28(4), 418-422 (2006)
Christophe Tastet et al.
Proteomics, 3(2), 111-121 (2003-02-26)
Several zwitterionic detergents differing in their polar heads, linker parts and hydrophobia tail were synthesized and evaluated for their efficiency in protein solubilizers for two-dimensional electrophoresis. A model system consisting of human red blood cell ghosts was used for this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service