Skip to Content
Merck
All Photos(1)

Key Documents

Y0000798

Pholcodine for peak identification

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Pholcodine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30N2O4
CAS Number:
Molecular Weight:
398.50
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

pholcodine

manufacturer/tradename

EDQM

drug control

regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C23H30N2O4/c1-24-7-6-23-16-3-4-18(26)22(23)29-21-19(5-2-15(20(21)23)14-17(16)24)28-13-10-25-8-11-27-12-9-25/h2-5,16-18,22,26H,6-14H2,1H3/t16?,17-,18+,22+,23+/m1/s1

InChI key

GPFAJKDEDBRFOS-SLITUTQKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Pholcodine for peak identification EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Florvaag et al.
Allergy, 61(1), 49-55 (2005-12-21)
A previous study showed a relation between pholcodine (PHO) consumption, prevalence of IgE-sensitization to PHO, morphine (MOR) and suxamethonium (SUX) and anaphylaxis to neuromuscular blocking agents (NMBA). The purpose of this pilot study was to explore the effect on IgE
IgE-mediated anaphylactic reactions to neuromuscular blocking agents: can they be prevented?
E Florvaag et al.
Current allergy and asthma reports, 8(5), 375-376 (2008-08-07)
Didier G Ebo et al.
Anesthesiology, 107(2), 253-259 (2007-08-02)
Diagnosis of allergy from neuromuscular blocking agents is not always straightforward. The objectives of the current study were to investigate the value of quantification of immunoglobulin E (IgE) by ImmunoCAP (Phadia AB, Uppsala, Sweden) in the diagnosis of rocuronium allergy
Allergy to pholcodine: first case documented by oral challenge.
F Codreanu et al.
Allergy, 60(4), 544-545 (2005-02-25)
Z Kovács et al.
Analytical and bioanalytical chemistry, 386(6), 1709-1716 (2006-09-28)
The acid-base properties of pholcodine, a cough-depressant agent, and related compounds including metabolites were studied by 1H NMR-pH titrations, and are characterised in terms of macroscopic and microscopic protonation constants. New N-methylated derivatives were also synthesized in order to quantitate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service