Skip to Content
Merck
All Photos(1)

Key Documents

NTSV

Sigma-Aldrich

Nerve Terminal Staining Kit V

Kit utilizes SynaptoRed C2 and the sulfonated β-cyclodextrin Advasep-7

Synonym(s):

Nerve Terminal Staining Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12171500

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

The Nerve Terminal Stain Kits are used to detect and study synaptic vesicle recycling in neuronal synapses and neuromuscular junctions. Although each kit employs a different amphiphilic styryl pyridinium dye to visualize the vesicles, the tissue preparation, reagents required, and staining procedures are identical for each. The dyes fluoresce intensely inside plasma membranes but only minimally in aqueous environments.
When neurons are actively releasing neurotransmitters, dye is incorporated into recycled vesicles at the presynaptic terminal. The dye-containing vesicles are not susceptible to subsequent wash steps that remove the excess dye from the outer leaflet of the plasma membrane. Thus only active presynaptic terminals are labeled. Advasep-7 further reduces background by encapsulating the probes, preventing stain from accumulating interstitially.

Kit Components Also Available Separately

Product No.
Description
SDS

  • S6689SynaptoRed C2, ≥95% (HPLC), solidSDS

  • A3723ADVASEP-7, solidSDS


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service