Skip to Content
Merck
All Photos(1)

Key Documents

H7148

Sigma-Aldrich

Hydroquinone

suitable for photographic applications, ≥99%

Synonym(s):

1,4-Benzenediol, 1,4-Dihydroxybenzene, HQ

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4-1,4-(OH)2
CAS Number:
Molecular Weight:
110.11
Beilstein:
605970
EC Number:
MDL number:
UNSPSC Code:
12352200

vapor density

3.81 (vs air)

vapor pressure

1 mmHg ( 132 °C)

Assay

≥99%

autoignition temp.

930 °F

bp

285 °C (lit.)

mp

172-175 °C (lit.)

suitability

suitable for photographic applications

SMILES string

Oc1ccc(O)cc1

InChI

1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H

InChI key

QIGBRXMKCJKVMJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1 - Muta. 2 - Skin Sens. 1B

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

329.0 °F - closed cup

Flash Point(C)

165 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Carbonnelle et al.
Toxicology and applied pharmacology, 132(2), 220-226 (1995-06-01)
Decreased interleukin-1 (IL-1) production by mononuclear phagocytes has been shown to contribute to benzene myelotoxicity in animals. The study presented here was designed to examine the relevance of this mechanism in humans. Fresh human blood monocytes were exposed to 0.001-10
Yaqi Hu et al.
Biosensors & bioelectronics, 47, 45-49 (2013-04-03)
A novel light "on-off" photoelectrochemical sensing for the sensitive determination of hydroquinone was developed based on the porphyrin-functionalized Au nanoparticles on graphene (porphyrin/AuNPs/graphene). Gold nanoparticles (AuNPs) were firstly modified onto graphene sheets using NaBH4 as reductant and the porphyrin/AuNPs/graphene nanocomposites
Jacob Levitt
Journal of the American Academy of Dermatology, 57(5), 854-872 (2007-05-01)
Recently, the US Food and Drug Administration proposed a ban on over-the-counter hydroquinone mainly on the basis of high absorption, reports of exogenous ochronosis in humans, and murine hepatic adenomas, renal adenomas, and leukemia with large doses over extended time
A P DeCaprio
Critical reviews in toxicology, 29(3), 283-330 (1999-06-24)
Hydroquinone (HQ) is a high-volume commodity chemical used as a reducing agent, antioxidant, polymerization inhibitor, and chemical intermediate. It is also used in over-the-counter (OTC) drugs as an ingredient in skin lighteners and is a natural ingredient in many plant-derived
Nuan P Cheah et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(3), 1072-1081 (2013-02-19)
Tobacco smoke consists of thousands of harmful components. A major class of chemicals found in tobacco smoke is formed by aldehydes, in particular formaldehyde, acetaldehyde and acrolein. The present study investigates the gene expression changes in human lung alveolar epithelial

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service