Skip to Content
Merck
All Photos(1)

Documents

71753

Sigma-Aldrich

Sodium 2-methyl-2-propanethiolate

technical, ≥90% (RT)

Synonym(s):

2-Methyl-2-propanethiol sodium salt, Sodium tert-butyl sulfide, t-Butanethiol sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSNa
CAS Number:
Molecular Weight:
112.17
Beilstein:
3682306
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥90% (RT)

SMILES string

CC(C)(C)S[Na]

InChI

1S/C4H10S.Na/c1-4(2,3)5;/h5H,1-3H3;/q;+1/p-1

InChI key

TUZFIWBNGKNFPW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Salts of alkanethiols are versatile intermediates for the synthesis of various thiol esters and thio ethers. In contrast to the stenchy gaseous or liquid thiols, the salts are easy to handle solids.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P. Cogolli et al.
The Journal of Organic Chemistry, 44, 2636-2636 (1979)
R.D. Little et al.
Journal of the American Chemical Society, 100, 4607-4607 (1978)
Shih-Ching Chuang et al.
The Journal of organic chemistry, 72(17), 6447-6453 (2007-07-20)
In the field of open-cage fullerenes, there was a lack of a universal standard that could correlate and quantify the orifice size of open-cage fullerenes. One cannot compare the relative orifice size by simple comparison of the number of atoms
Exploration of 9-bromo [7] helicene reactivity.
Zadny J, et al.
Tetrahedron, 69(3), 6213-6218 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service