Skip to Content
Merck
All Photos(1)

Documents

67123

Millipore

Preston Campylobacter growth supplement

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171614
NACRES:
NA.74

shelf life

limited shelf life, expiry date on the label

Quality Level

application(s)

microbiology

storage temp.

2-8°C

Application

A chemical enrichment recommended for the enhancement of growth and aerotolerance of Campylobacter spp..

Components

Composition:
Per vial sufficient for 500 ml medium
Ingredients Concentration
Sodium pyruvate 0.125g
Sodium metabisulfite 0.125g
Ferrous sulfate 0.125g

Reconstitution

Directions:
Rehydrate the contents of 1 vial aseptically with 2 ml of sterile distilled water and mix well to dissolve. Avoid frothing of the solution. Aseptically add the contents to 500 ml of sterile, molten, cooled (45-50°C) Columbia Blood Agar Base, Blood Agar Base, Brucella Agar Base or Preston Broth. Mix gently and pour into sterile petri plates or tubes.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - Skin Sens. 1

Supplementary Hazards

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service