Skip to Content
Merck
All Photos(1)

Key Documents

63195

Supelco

Maize powder MZ 0, IRMM certified reference material No. 411

BioChemika, for the detection of GMO maize (Bt-176) with PCR

Synonym(s):

Corn flour MZ 0, IRMM certified reference material, GMO maize powder MZ 0 (0% Bt-176)

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$86.80

$86.80


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
2 G
$86.80

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$86.80


Estimated to ship on23 April 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

BioChemika

quality

for the detection of GMO maize (Bt-176) with PCR

composition

genetically modified Bt-176 maize, 0%

application(s)

microbiology
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service