Skip to Content
Merck
All Photos(1)

Documents

59340

Sigma-Aldrich

2-(Isopropylamino)ethanol

technical, ≥95% (GC)

Synonym(s):

N-Isopropylethanolamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHNHCH2CH2OH
CAS Number:
Molecular Weight:
103.16
Beilstein:
1633453
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

technical

Assay

≥95% (GC)

refractive index

n20/D 1.441 (lit.)
n20/D 1.441

bp

172 °C (lit.)

density

0.897 g/mL at 25 °C (lit.)

SMILES string

CC(C)NCCO

InChI

1S/C5H13NO/c1-5(2)6-3-4-7/h5-7H,3-4H2,1-2H3

InChI key

RILLZYSZSDGYGV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service