Skip to Content
Merck
All Photos(1)

Key Documents

52495

Sigma-Aldrich

Hexadimethrine bromide

for protein sequence analysis, ≥94% (AT)

Synonym(s):

1,5-Dimethyl-1,5-diazaundecamethylene polymethobromide, Polybrene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200

grade

for protein sequence analysis

Assay

≥94% (AT)

solubility

H2O: 0.1 g/mL, clear to slightly hazy, colorless to faintly yellow

storage temp.

2-8°C

InChI

1S/C10H24N2.C3H6Br2/c1-11(2)9-7-5-6-8-10-12(3)4;4-2-1-3-5/h5-10H2,1-4H3;1-3H2

InChI key

KZKAYEGOIJEWQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Facilitates automated sequencing of small peptides.[1] Used to coat glass microfiber sheets for protein blotting.[2]

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Vandekerckhove et al.
European journal of biochemistry, 152(1), 9-19 (1985-10-01)
A procedure has been developed which allows the immobilization on glass-fiber sheets coated with the polyquaternary amine, Polybrene, of proteins and protein fragments previously separated on sodium-dodecylsulfate-containing polyacrylamide gels. The transfer is carried out essentially as has been used for
Automated amino acid sequence of small peptides utilizing Polybrene.
D G Klapper et al.
Analytical biochemistry, 85(1), 126-131 (1978-03-01)
Kimberly A Brown et al.
Journal of cellular biochemistry, 105(2), 596-611 (2008-08-30)
Transforming growth factor-beta 1 (TGF-beta1) is an important growth inhibitor of epithelial cells and insensitivity to this cytokine results in uncontrolled cell proliferation and can contribute to tumorigenesis. TGF-beta1 signals through the TGF-beta type I and type II receptors, and
Hua Yang et al.
Investigative ophthalmology & visual science, 51(1), 28-34 (2009-08-08)
Pigment epithelium-derived factor (PEDF) is known to be an angiogenesis suppressor and to have antitumor effects. This study investigates whether constitutive overexpression of PEDF inhibits the growth and hepatic micrometastasis of ocular melanoma. Real-time RT-PCR was used to detect endogenous
[Chemiluminescence of peritoneal macrophages activated by non-natural polyelectrolytes].
L G Korkina et al.
Doklady Akademii nauk SSSR, 282(1), 206-209 (1985-05-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service