522082
Trichloroacetic acid
biotech. grade, redistilled, ≥99%
Synonym(s):
TCA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
biotech. grade
vapor density
<1 (vs air)
vapor pressure
1 mmHg ( 51 °C)
Assay
≥99%
refractive index
n20/D 1.62 (lit.)
bp
196 °C (lit.)
mp
54-58 °C (lit.)
density
1.62 g/mL at 25 °C (lit.)
SMILES string
OC(=O)C(Cl)(Cl)Cl
InChI
1S/C2HCl3O2/c3-2(4,5)1(6)7/h(H,6,7)
InChI key
YNJBWRMUSHSURL-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Suitable for protein sequencing
Traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. Used as decalcifier and fixative in microscopy.
Other Notes
A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1A
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
>235.4 °F - closed cup
Flash Point(C)
> 113 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Trichloroacetic acid peels.
Dermatologic Therapy, 13(2), 173-182 (2000)
Trichloroacetic acid at 105 K.
Acta Crystallographica Section E, Structure Reports Online, 59(3), 316-318 (2003)
Toxicology and applied pharmacology, 90(2), 183-189 (1987-09-15)
Trichloroethylene (TCE) has previously been shown to be carcinogenic in mouse liver when administered by daily gavage in corn oil. The metabolism of TCE results, in part, in the formation of trichloroacetic acid (TCA) as a major metabolite and dichloroacetic
Cancer discovery, 4(9), 1014-1021 (2014-06-18)
Metastatic solid tumors are almost invariably fatal. Patients with disseminated small-cell cancers have a particularly unfavorable prognosis, with most succumbing to their disease within two years. Here, we report on the genetic and functional analysis of an outlier curative response
Environment international, 30(8), 1119-1150 (2004-09-01)
Trichloroacetic acid (TCAA) is a member of the family of compounds known as chloroacetic acids, which includes mono-, di- and trichloroacetic acid. The significant property these compounds share is that they are all phytotoxic. TCAA once was widely used as
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service