Skip to Content
Merck
All Photos(1)

Key Documents

49902

Supelco

Tin(II) ionophore I

Selectophore

Synonym(s):

Dibenzo-18-crown-6, 2,3,11,12-Dibenzo-1,4,7,10,13,16-hexaoxacyclooctadeca-2,11-diene, 6,7,9,10,17,18,20,21-Octahydrodibenzo[b,k][1,4,7,10,13,16]hexaoxacyclooctadecene, DB18C6

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O6
CAS Number:
Molecular Weight:
360.40
Beilstein:
1162153
EC Number:
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

mp

162-164 °C (lit.)

SMILES string

C1COc2ccccc2OCCOCCOc3ccccc3OCCO1

InChI

1S/C20H24O6/c1-2-6-18-17(5-1)23-13-9-21-11-15-25-19-7-3-4-8-20(19)26-16-12-22-10-14-24-18/h1-8H,9-16H2

InChI key

YSSSPARMOAYJTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Visit our Sensor Applications portal to learn more.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoyuki Akutagawa et al.
Chemistry, an Asian journal, 5(3), 520-529 (2009-12-24)
The supramolecular (HOPD(+))(DB[18]crown-6) cation, in which HOPD(+) and DB[18]crown-6 are o-aminoanilinium and dibenzo[18]crown-6, respectively, was introduced into [Ni(dmit)(2)](-) salts (dmit(2-) = 2-thioxo-1,3-dithiole-4,5-dithiolate). Conformational polymorphs were observed as tetragonal (HOPD(+))(DB[18]crown-6)[Ni(dmit)(2)](-) (1) and monoclinic (HOPD(+))(DB[18]crown-6)[Ni(dmit)(2)](-) (2). The ammonium group of HOPD(+) in
Chang Min Choi et al.
The journal of physical chemistry. A, 114(42), 11167-11174 (2010-10-22)
Ultraviolet photodepletion spectra of dibenzo-18-crown-6-ether complexes with alkaline earth metal divalent cations (A(2+)-DB18C6, A = Ba, Sr, Ca, and Mg) were obtained in the gas phase using electrospray ionization quadrupole ion-trap reflectron time-of-flight mass spectrometry. Each spectrum exhibits the lowest
Marcin Dułak et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(2), 532-548 (2006-01-03)
Experimental (IR and Raman) and theoretical (Kohn-Sham calculations) methods are used in a combined analysis aimed at refining the available structural data concerning the molecular guests in channels formed by stacked dibenzo-18-crown-6 (DB18C6) crown ether. The calculations are performed for
Toru Endo et al.
Dalton transactions (Cambridge, England : 2003), 40(7), 1491-1496 (2011-01-19)
A supramolecular cation of (m-FAni(+))(DB[18]crown-6), where m-FAni(+) and DB[18]crown-6 denote m-fluoroanilinium(+) and dibenzo[18]crown-6, respectively, which is the polar unit rotating in the ferroelectric crystal of (m-FAni(+))(DB[18]crown-6)[Ni(dmit)(2)](-), was introduced into a ferromagnetic [MnCr(oxalate)(3)](-) salt as the counter cation. The crystal structure
Lei Hu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 104, 87-91 (2012-12-26)
In this contribution, an improved near-infrared absorbing colorimetric detection of Hg(2+) ion with high selectivity and sensitivity had been developed by using bis-dibenzo-18-crown-6-based squaraine (BCSQ) as sensor in aqueous media. The proposed method was applied to analyze trace Hg(2+) in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service