Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

47657

Sigma-Aldrich

Fmoc-Gln(Dod)-Wang resin

extent of labeling: 0.4-0.6 mmol/g loading

Synonym(s):

Nα-Fmoc-Nδ-(4,4′-dimethoxybenzhydryl)-L-glutamine 4-benzyloxybenzyl ester polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Pricing and availability is not currently available.

extent of labeling

0.4-0.6 mmol/g loading

matrix

polystyrene, crosslinked with 1% DVB

particle size

200-400 mesh

storage temp.

2-8°C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service