Skip to Content
Merck
All Photos(1)

Documents

36162

Supelco

Metobromuron

PESTANAL®, analytical standard

Synonym(s):

N′-(4-Bromophenyl)-N-methoxy-N-methylurea

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H11BrN2O2
CAS Number:
Molecular Weight:
259.10
Beilstein:
2103964
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CON(C)C(=O)Nc1ccc(Br)cc1

InChI

1S/C9H11BrN2O2/c1-12(14-2)9(13)11-8-5-3-7(10)4-6-8/h3-6H,1-2H3,(H,11,13)

InChI key

WLFDQEVORAMCIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Metobromuron is a phenylurea herbicide widely used in pre- and post-emergence control of blackgrass, silky bentgrass, wild oats and annual meadow grass.

Application

Metobromuron may be used as an analytical reference standard for the quantification of the analyte in vegetables, soil, tobacco and aqueous samples by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The determination of some substituted urea herbicide residues in soil by electron-capture gas chromatography.
McKone CE
Journal of Chromatography A, 44(1-2), 60-66 (1969)
Method development for the determination of selected pesticides on tobacco by high-performance liquid chromatography--electrospray ionisation-tandem mass spectrometry.
Mayer-Helm B, et al.
Talanta, 74(5), 1184-1190 (2008)
Application of solid-phase microextraction for determining phenylurea herbicides and their homologous anilines from vegetables.
Berrada H, et al.
Journal of Chromatography A, 1042(1-2), 9-14 (2004)
B Anić et al.
Arhiv za higijenu rada i toksikologiju, 50(2), 193-199 (1999-11-24)
An 81-year-old man was admitted in the emergency department approximately four hours after accidental, ingestion of an unknown quantity of herbicide "Galex 500 EC". This product contains 25% of metolachlor and 25% of metobromuron dissolved in xylene. In spite of
Vincent J Smith et al.
Carbohydrate research, 344(17), 2388-2393 (2009-10-06)
We report the formation of inclusion complexes between the phenylurea herbicide metobromuron [3-(p-bromophenyl)-1-methoxy-1-methylurea] and beta- and gamma-cyclodextrin in the solid state. Formation of crystalline inclusion complexes by the kneading method was confirmed by powder X-ray diffraction and further structural characterization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service