Skip to Content
Merck
All Photos(1)

Key Documents

16186

Sigma-Aldrich

SODOSIL RAM 05

acid cleaner with formic acid

Sign Into View Organizational & Contract Pricing


About This Item

UBA Number:
08050021
UNSPSC Code:
12352100

pH

1-3 (20 °C, 1%)

density

1.06-1.12 g/mL at 20 °C

Application

SODOSIL was used in cleaning plastic ware during determination of lead in whole blood specimens[1]. It was used in rinsing epoxy-coated surfaces[2], multielectrode arrays[3] and mineral surfaces[4].

Reconstitution

The best cleaning results are achieved with = 10−20% for normal contamination. The cleaning time = 2−24 h depends on the degree of contamination and on the bath temperature. The action of ultra-sonic waves accelerates the cleaning operation considerably as does an increase in temperature. The cleaning solution can be prepared with tap water. The use of demineralisied water results in more optimum utilisation of baths.

Legal Information

SODOSIL is a trademark of Sigma-Aldrich Chemie GmbH

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachary M Aman et al.
Langmuir : the ACS journal of surfaces and colloids, 29(50), 15551-15557 (2013-11-26)
Clathrate hydrate adhesion forces play a critical role in describing aggregation and deposition behavior in conventional energy production and transportation. This manuscript uses a unique micromechanical force apparatus to measure the adhesion force between cyclopentane hydrate and heterogeneous quartz and
Shimshon Jacobi et al.
Journal of neurophysiology, 97(5), 3597-3606 (2007-03-09)
We investigate the propagation of neural activity along one-dimensional rat hippocampal cultures patterned in lines over multielectrode arrays. Activity occurs spontaneously or is evoked by local electrical or chemical stimuli, with different resulting propagation velocities and firing rate amplitudes. A
G Aspenes et al.
Journal of colloid and interface science, 343(2), 529-536 (2009-12-29)
The mechanisms by which hydrates deposit in a petroleum production line are related to pipeline surface properties, fluid composition and properties, and water cut. In this work, adhesion forces between cyclopentane hydrates and solid surfaces were investigated as a function
Determination of lead in whole blood by graphite furnace atomic absorption spectrometry with matrix modification.
Navarro JA, et al.
Journal of Analytical Atomic Spectrometry, 4(5), 401-406 (1989)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service