06914
Thiophene
analytical standard
Synonym(s):
Thiofuran, Thiole
About This Item
Recommended Products
grade
analytical standard
Quality Level
vapor density
2.9 (vs air)
vapor pressure
40 mmHg ( 12.5 °C)
Assay
≥99.5% (GC)
autoignition temp.
743 °F
shelf life
limited shelf life, expiry date on the label
expl. lim.
12.5 %
technique(s)
HPLC: suitable
gas chromatography (GC): suitable
impurities
~0.2% benzene
refractive index
n20/D 1.529 (lit.)
n20/D 1.529
bp
84 °C (lit.)
mp
−38 °C (lit.)
density
1.051 g/mL at 25 °C (lit.)
application(s)
environmental
format
neat
SMILES string
c1ccsc1
InChI
1S/C4H4S/c1-2-4-5-3-1/h1-4H
InChI key
YTPLMLYBLZKORZ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 3 - Carc. 1A - Eye Irrit. 2 - Flam. Liq. 2 - Muta. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
15.8 °F
Flash Point(C)
-9 °C
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service