Skip to Content
Merck
All Photos(1)

Key Documents

17-0080

Sigma-Aldrich

Kerosene

SAJ special grade, ≥90.0%

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
15101502
grade:
SAJ special grade
Assay:
≥90.0%
bp:
190-250 °C (lit.)
vapor pressure:
0.23 mmHg ( 20 °C)

grade

SAJ special grade

vapor density

4.5 (vs air)

vapor pressure

0.23 mmHg ( 20 °C)

Assay

≥90.0%

form

liquid

autoignition temp.

442 °F

expl. lim.

5 %

availability

available only in Japan

bp

190-250 °C (lit.)

density

0.8 g/mL at 25 °C (lit.)

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

179.6 °F - closed cup

Flash Point(C)

82 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kent L Gee et al.
The Journal of the Acoustical Society of America, 123(6), 4082-4093 (2008-06-10)
To address the question of the role of nonlinear effects in the propagation of noise radiated by high-power jet aircraft, extensive measurements were made of the F-22A Raptor during static engine run-ups. Data were acquired at low-, intermediate-, and high-thrust
Gerardo Ramos et al.
Toxicology and applied pharmacology, 195(3), 331-338 (2004-03-17)
Applying military jet fuel (JP-8) or commercial jet fuel (Jet-A) to the skin of mice suppresses the immune response in a dose-dependent manner. The release of biological response modifiers, particularly prostaglandin E2 (PGE2), is a critical step in activating immune
J A Namocatcat et al.
Journal of contaminant hydrology, 67(1-4), 177-194 (2003-11-11)
Evolution of trimethylbenzoic acids in the KC-135 aquifer at the former Wurtsmith Air Force Base (WAFB), Oscoda, MI was examined to determine the functionality of trimethylbenzoic acids as key metabolite signatures in the biogeochemical evolution of an aquifer contaminated with
J Rossi et al.
Journal of toxicology and environmental health. Part A, 63(6), 397-428 (2001-08-03)
The U.S. Naval Service is anticipating transition from the nearly exclusive use of JP-5 jet fuel to predominant use of JP-8, consistent with the primary utilization by the U.S. Army, U.S. Air Force, and the militaries of most NATO countries.
Narayanasamy Kanikkannan et al.
Toxicology, 175(1-3), 35-47 (2002-06-07)
Jet A and JP-8 are the major jet fuels used in civilian and military (US Air Force) flights, respectively. JP-8+100 is a new jet fuel recently introduced by US Air Force in some of its locations. The purpose of this

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service