Skip to Content
Merck
All Photos(1)

Key Documents

S7907-M

Sigma-Aldrich

Amplifluor® SNPs Genotyping System for Assay Development

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116012
eCl@ss:
32160405
NACRES:
NA.84

application(s)

genomic analysis

Quality Level

detection method

fluorometric

General description

The Amplifluor SNPs Genotyping System for Assay Development is designed for new users unfamiliar with the technology. This kit contains the fluorescein-, sulforhodamine- and JOE-labeled Amplifluor UniPrimers as well as control primers and templates.

Legal Information

Amplifluor is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service