Skip to Content
Merck
All Photos(3)

Key Documents

MIACFD201

Millipore

Microfil® Funnel Dispenser

For use with 250 mL Microfil® funnels

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
$1,640.00

$1,640.00


Estimated to ship on23 April 2025


Request more information

Select a Size

Change View
1 EA
$1,640.00

About This Item

UNSPSC Code:
41116129
NACRES:
NB.24

$1,640.00


Estimated to ship on23 April 2025


Request more information

Quality Level

packaging

pkg of 1 unit

manufacturer/tradename

Microfil® with S-Pak®

input

sample type beverage(s)
sample type food(s)

application(s)

bioburden testing
clinical
environmental
food and beverages
industrial qc

compatibility

for use with EZ-Fit® Filtration Manifold

shipped in

ambient

General description

The funnel dispenser for the 250 mL funnel is an additional accessory of the Microfil® filtration system. The Microfil® filtration system is a simple and reliable solution for routine microbiological testing. The push-fit design of the Microfil® funnels seals tightly to manifold supports without the need for clamps or O-rings. The funnels, packed in convenient packs of 25, are loaded to the custom-designed funnel dispenser and can be easily removed as needed. The funnel dispenser is designed to allow rapid dispensing of Microfil® funnels without compromising sterility.

Application

The funnel dispenser is an additional accessory of the Microfil® filtration system used in general microbiological analysis of beer, bottled water, pharmaceutical products analysis, environmental analysis, bioburden testing and water monitoring.

Legal Information

EZ-FIT is a registered trademark of Merck KGaA, Darmstadt, Germany
MICROFIL is a registered trademark of Merck KGaA, Darmstadt, Germany
S-PAK is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Microfil® filtration system ensures microbiological quality assurance in routine testing of raw materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service