Skip to Content
Merck
All Photos(4)

Documents

MABS1295

Sigma-Aldrich

Anti-mS2i6A Antibody, clone CL-3

clone CL-3, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

CL-3, monoclonal

species reactivity (predicted by homology)

mammals (cells containing mitochondria)

packaging

antibody small pack of 25 μg

technique(s)

ELISA: suitable
dot blot: suitable
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG2aκ

shipped in

ambient

target post-translational modification

unmodified

General description

2-methylthio-N6-isopentenyladenosine (mS2i6A) is a hypermodified modified nucleoside that has been shown to be present in mitochondrial tRNA. It reads codons starting with uridine. The mS2 group of mS2i6A is critical for the accurate decoding of Tyr and Phe codons. Higher concentrations of mS2i6A are found in tissues with a high energy demand, such as muscle and brain. One of the major role of mS2i6A is to stabilize the stacking interactions of the anticodon loop helix and thereby prevent codon misreading. Its deficiency has been linked to a higher rejection rate of tRNA4Leu by more aggressive proofreading on the wild-type ribosome. However, it is shown that mS2i6A has no effect on codon-anticodon interactions on wild-type ribosomes as long as aminoacyl-tRNA is in ternary complex with EF-Tu and GTP. Also, it has been reported that the lack of mS2i6A does not decrease the ability of the tRNA to be charged with its cognate amino acid, but it alters the efficiency with which the tRNA is bound to the ribosome. CDK5RAP1, which inhibits the activity of CDK5, is shown to be an N6-isopentenyladenosine methylthiotransferase. It converts the RNA modification N6-isopentenyladenosine (i6A) in mS2i6A post-synthetically. This conversion is observed both in the mitochondrial as well as nuclear RNA species. (Ref.: Wilson, RK and Roe, BA (1989). Proc. Natl. Acad. Sci. USA. 86(2); 409-413, Diaz, I., and Ehrenberg, M (1991). J. Mol. Biol. 222(4); 1161-71; Reiter, V et al. (2012). Nucleic Acids Res. 40(13); 6235-40).

Specificity

Clone 3 (CL-3) recognizes wild-type total RNA but not knockout total RNA.

Immunogen

A synthesized mS2i6A oligonucleotide conjugated to BSA.

Application

Anti-MS2i6A, clone CL-3, Cat. No. MABS1295, is a mouse mouse monoclonal antibody that detects mS2i6A and has been tested for use in Dot Blot, ELISA, Immunocytochemistry, and Immunofluorescence.
ELISA Analysis: A representative lot detected mS2i6A in ms2i6A or i6A wells (Courtesy of Fan-Yan Wei, Ph.D., Faculty of Life Sciences, Kumamoto University Japan).

Immunofluorescence Analysis: A representative lot detected mS2i6A in HeLa cells treated with mitotracker (Courtesy of Fan-Yan Wei, Ph.D., Faculty of Life Sciences, Kumamoto University Japan).

Dot Blot Analysis: A 1:200 dilution from a representative lot detected mS2i6A in Wild type (WT) total RNA and Cdk5rap1 KO mice total RNA samples from liver (Courtesy of Fan-Yan Wei, Ph.D., Faculty of Life Sciences, Kumamoto University Japan).

Quality

Evaluated by Immunocytochemistry in HeLa cells with mitotracker.

Immunocytochemistry Analysis: A 1:500 dilution of this antibody detected mS2i6A in HeLa cells with Mitotracker.

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service