Skip to Content
Merck
All Photos(2)

Documents

MABF856

Sigma-Aldrich

Anti-Helios Antibody, clone 22F6

clone 22F6, from hamster(Armenian)

Synonym(s):

Zinc finger protein Helios, Ikaros family zinc finger protein 2, Helios

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

hamster (Armenian)

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

22F6, monoclonal

species reactivity

human, mouse

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IKZF2(22807)

Related Categories

General description

Zinc finger protein Helios (Uniprot Q9UKS7; also known as Ikaros family zinc finger protein 2, Zinc finger DNA binding protein Helios, Zinc finger protein subfamily 1A member 2) is encoded by the IKZF2 (also known as ANF1A2, HELIOS, ZNF1A2, ZNFN1A2) gene (Gene ID 22807) in human. Helios, Ikaros, Aiolos, Eos, and Pegasus constitute a family of transcription factors characterized by six highly conserved C2H2 zinc fingers. Their four N-terminal zinc fingers mediate DNA binding, while the two C-terminal zinc fingers are responsible for homodimeric and heterodimeric protein interactions. Eos and Pegasus are expressed in a wide variety of tissues, whereas Helios, Ikaros, and Aiolos are limited to the hematopoietic system. Specifically, Helios expression is reported to be restricted to a subpopulation (approximately 70%) of Foxp3(+) T cells in mice and humans.

Specificity

Expected to react with isoform 1, 2 (Helios 1v), 3 (Helios del(Ex3,4)), 4 (Helios del(Ex6)v), 5 (Helios 1+2a,2b, Helios 1v+2a, Helios del(Ex3)+2a), 6 (Helios 1+5a, Helios 1+5a,5b), 7 (Helios del(Ex5)v), but not isoform 8 (Helios S).

Immunogen

Epitope: N-terminal
Linear peptide corresponding to a human Helios N-terminal sequence.

Application

Flow Cytometry Analysis: 1.0 µg of this antibody from a representative lot detected Helios in 1X10E6 human Jurkat cells.
Flow Cytometry Analysis: A representative lot detected Helios immunoreactivity among the majority of CXCR5-positive cells within the CD4+Foxp3+ population of murine peripheral lymph node (LN) cells (Chung, Y., et al. (2011). Nat Med.17(8):983-988).
Flow Cytometry Analysis: A representative lot detected Helios immunoreactivity in the Foxp3-positive, but not Foxp3-negative, subpopulation of CD4+ cells from murine peripheral lymph node (LN) and human PBMCs (Thornton, A.M., et al. (2010). J Immunol.184(7):3433-3441).
Flow Cytometry Analysis: A representative lot revealed the age dependency of Helios-positive population among CD4+ murine splenocytes and CD4+/CD8- murine thymocytes (Thornton, A.M., et al. (2010). J Immunol.184(7):3433-3441).
Flow Cytometry Analysis: A representative lot detected increased Helios-positive population among the TGF-beta-stimulated Treg, but not Teff cells from murine peripheral lymph node (LN) and human PBMCs (Thornton, A.M., et al. (2010). J Immunol.184(7):3433-3441).
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Anti-Helios Antibody, clone 22F6 is validated for use in Flow Cytometry for the detection of Helios.

Quality

Evaluated by Flow Cytometry in human PBMCs.

Flow Cytometry Analysis: 1.0 µg of this antibody detected Helios in human PBMCs.

Target description

~ 59 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified Armenian hamster monoclonal IgG antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service