Skip to Content
Merck
All Photos(2)

Documents

MABE985

Sigma-Aldrich

Anti-Fox1 Antibody, clone 1D10

clone 1D10, from mouse

Synonym(s):

RNA binding protein fox-1 homolog 1, Ataxin-2-binding protein 1, Fox-1 homolog A, Fox1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D10, monoclonal

species reactivity

mouse

technique(s)

RIP: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Rbfox3(52897)

General description

RNA binding protein fox-1 homolog 1 (UniProt Q9JJ43; also known as Ataxin-2-binding protein 1, Fox-1 homolog A, Hexaribonucleotide binding protein 1) is encoded by the Rbfox1 (also known as A2bp, A2bp1, Fox-1, Hrnbp1) gene (ORF MNCb-3035; Gene ID 268859) in murine species. The RNA-binding Fox (Rbfox) family of splicing factors is comprised of three members, Rbfox1 (Fox-1 or A2BP1), Rbfox2 (Fox-2 or RBM9), and Rbfox3 (Fox-3, HRNBP3 or NeuN). Rbfox proteins regulate splicing of many neuronal transcripts (pre-mRNAs) by binding the sequence (U)GCAUG in introns flanking alternative exons. A (U)GCAUG motif that lies downstream of the alternative exon generally promotes Rbfox-dependent exon inclusion, whereas an upstream motif will usually repress exon inclusion. Rbfox1 transcript itself is subjected to alternative splicing in response to chronic cell depolarization. This results in the increased expression of the nuclear, splicing-active Rbfox1 isoform, which in turn leads to increased splicing of Rbfox1 target transcripts of proteins, including N-methyl D-aspartate (NMDA) receptor 1 (Grin1) and a calcium ATPase (Atp2b1), that play important roles in neuronal excitation and calcium homeostasis.

Immunogen

GST-tagged recombinant protein corresponding to mouse Fox1.

Application

Anti-Fox1 Antibody, clone 1D10, Cat. No. MABE985, is a highly specific mouse Monoclonal antibody, that targets RNA binding protein fox-1 homolog 1 and has been tested in Western Blotting, Immunocytochemistry, Immunohistochemistry and RNA-Binding Immunoprecipitation/iCLIP.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Nuclear Receptors
Western Blotting Analysis: 0.5 µg/mL from a representative lot detected Fox1 in 10 µg of adult mouse frontal cortex tissue lysate.
Immunohistochemistry Analysis: A representative lot detected Fox1 in coronal sections from E18 mouse (Tang, Z.Z., et al. (2009). Mol Cell Biol. 29(17):4757-4765).
Immunocytochemistry Analysis: A representative lot detected Fox1 in post-mitotic neuronal cells differentiated from mouse embryonal carcinoma P19 cells (Lee, J.A., et al. (2009). Genes Dev. 23(19):2284-2293).
RNA-Binding Protein Immunoprecipitation/iCLIP Analysis: A representative lot was employed to immunoprecipitate Fox1-associated pre-mRNAs from UV-crosslinked murine cortex homogenates by individual-nucleotide resolution crosslinking immunoprecipitation (iCLIP) for characterization of Fox1 pre-mRNA-binding sites (Gehman, L.T., et al. (2011) Nat Genet. 43(7):706-711).
Western Blotting Analysis: A representative lot detected Fox1 in mouse cortical tissue homogenate (Tang, Z.Z., et al. (2009). Mol Cell Biol. 29(17):4757-4765).

Quality

Evaluated by Western Blotting in adult mouse brain tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected Fox1 in 10 µg of adult mouse brain tissue lysate.

Target description

~43 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2aκ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sebastien M Weyn-Vanhentenryck et al.
Nature communications, 9(1), 2189-2189 (2018-06-08)
Alternative splicing (AS) is one crucial step of gene expression that must be tightly regulated during neurodevelopment. However, the precise timing of developmental splicing switches and the underlying regulatory mechanisms are poorly understood. Here we systematically analyze the temporal regulation
Brie Wamsley et al.
Neuron, 100(4), 846-859 (2018-10-16)
Cortical interneurons display a remarkable diversity in their morphology, physiological properties, and connectivity. Elucidating the molecular determinants underlying this heterogeneity is essential for understanding interneuron development and function. We discovered that alternative splicing differentially regulates the integration of somatostatin- and
Ainara Elorza et al.
Brain : a journal of neurology, 144(7), 2009-2023 (2021-03-17)
Correction of mis-splicing events is a growing therapeutic approach for neurological diseases such as spinal muscular atrophy or neuronal ceroid lipofuscinosis 7, which are caused by splicing-affecting mutations. Mis-spliced effector genes that do not harbour mutations are also good candidate
Francesco Tomassoni-Ardori et al.
eLife, 8 (2019-08-21)
Brain-derived neurotrophic factor (BDNF) is a potent modulator of brain synaptic plasticity. Signaling defects caused by dysregulation of its Ntrk2 (TrkB) kinase (TrkB.FL) and truncated receptors (TrkB.T1) have been linked to the pathophysiology of several neurological and neurodegenerative disorders. We
Francesco Tomassoni-Ardori et al.
Bio-protocol, 10(15) (2020-09-29)
Primary culture of mouse hippocampal neurons is a very useful in vitro model for studying neuronal development, axonal and dendritic morphology, synaptic functions, and many other neuronal features. Here we describe a step-by-step process of generating primary neurons from mouse

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service