Skip to Content
Merck
All Photos(2)

Key Documents

MABC140

Sigma-Aldrich

Anti-ARC Antibody, clone 6F9.1

clone 6F9.1, from mouse

Synonym(s):

Activity-regulated cytoskeleton-associated protein, ARC/ARG3.1, Activity-regulated gene 3.1 protein homolog, Arg3.1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$803.00

$803.00


Estimated to ship on26 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$803.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$803.00


Estimated to ship on26 April 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6F9.1, monoclonal

species reactivity

human

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ARC(23237)

General description

Activity-regulated cytoskeleton-associated protein (ARC), also known as Activity-regulated gene 3.1 protein homolog (ARG3.1), plays an important role in synaptic plasticity and the formation of long-term memory. ARC is enriched in the postsynaptic density of dendritic spines and regulates endocytosis of AMPA receptors in response to synaptic activity and is required for homeostatic synaptic scaling of AMPA receptors. ARC also plays a role in the regulation of cell morphology, cytoskeletal organization, stress fiber dynamics, and cell migration.

Immunogen

GST-tagged recombinant protein corresponding to human ARC.

Application

Anti-ARC Antibody, clone 6F9.1 is a highly specific mouse monoclonal antibody, that targets ARC & has been tested in western blotting & IHC (Paraffin).
Immunohistochemistry Analysis: A 1:1,000 dilution from a representative lot detected ARC in human brain tissue.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional

Quality

Evaluated by Western Blotting in human hippocampus tissue lysate.

Western Blotting Analysis: 0.2 µg/mL of this antibody detected ARC in 200 µg of human hippocampus tissue lysate.

Target description

~ 45 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Human hippocampus tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service