Skip to Content
Merck
All Photos(2)

Documents

MABC1137

Sigma-Aldrich

Anti-RKIP Antibody, clone 4A11

clone 4A11, from mouse

Synonym(s):

Raf kinase inhibitor protein, Phosphatidylethanolamine-binding protein 1, PEBP-1, HCNPpp, Neuropolypeptide h3, Prostatic-binding protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

purified antibody

antibody product type

primary antibodies

clone

4A11, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μg

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... PEBP1(5037)

General description

Phosphatidylethanolamine-binding protein 1 (UniProt: P30086; also known as PEBP-1, HCNPpp, Neuropolypeptide h3, Prostatic-binding protein, Raf kinase inhibitor protein, RKIP) is encoded by the PEBP1 (also known as PBP, PEBP) gene (Gene ID: 5037) in human. RKIP is as member of the phosphatidylethanolamine-binding-protein (PEBP) family that antagonizes multiple cell-survival pathways, such as the Ras-Raf-1, MEK/ERK, NF- B, and G-protein-coupled receptor Kinase 2 and thereby modulates cell growth, apoptosis, motility, invasion and metastasis. Over-expression of RKIP is shown to inhibit c-Src auto-phosphorylation and activation and IL-6-, JAK1 and 2-, and Raf-mediated STAT3 phosphorylation and activation. RKIP overexpression is shown to sensitize tumor cells to chemotherapeutic drug-induced apoptosis. Hence, it is considered as a suppressor of metastasis and a prognostic marker in several cancers. Clone 4A11 exhibits specific staining in formalin-fixed NIH3T3 cells transfected with GFP-PEBP1 and in some pancreatic cancer cells. (Ref.: Odabaei, G., et al. (2004). Adv. Cancer Res. 91; 169-200; Yousuf, S., et al. (2014). PLoS One 9(3); e92478).

Specificity

Clone 4A11 specifically detects human Raf kinase inhibitor protein, (RKIP).

Immunogen

Purified native and denatured Raf kinase inhibitor protein (RKIP).

Application

Detects Raf kinase inhibitor protein (Phosphatidylethanolamine-binding protein 1) using this mouse monoclonal Anti-RKIP, clone 4A11, Cat. No. MABC1137, tested for use in Western Blotting, Immunocytochemistry, and Immunohistochemistry (Paraffin).
Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected RKIP in human pancreas and human thyroid tissue sections.

Immunohistochemistry Analysis: A representative lot of this antibody detected RKIP in Immunohistochemistry applications (Wang, X., et. al. (2010). J Immunol Methods. 362(1-2):151-60).

Immunocytochemistry Analysis: A representative lot of this antibody detected RKIP in Immunocytochemistry applications (Wang, X., et. al. (2010). J Immunol Methods. 362(1-2):151-60).
Research Category
Apoptosis & Cancer

Quality

Evaluated by Western Blotting in HEK293 cell lysate.

Western Blotting Analysis: 1 µg/mL of this antibody detected RKIP in HEK293 cell lysate.

Target description

~23 kDa; 21.06 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein G purified
Purified mouse monoclonal antibodyIgG1κ in buffer containing PBS without azide

Storage and Stability

Stable for 1 year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service