Skip to Content
Merck
All Photos(1)

Documents

MAB5226

Sigma-Aldrich

Anti-Amyloid Antibody, β 1-40, clone OM16

clone OM16, Chemicon®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

OM16, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... APP(351)

Specificity

This antibody reacts with Amyloid-Beta protein 1-40 (bAP1-40) and secretory amyloid precursor protein (sAPP), but not with Amyloid-Beta protein 17-42.

Immunogen

Synthetic peptide of Amyloid-Beta protein 1-40.

Application

Anti-Amyloid Antibody, β 1-40, clone OM16 detects level of Amyloid & has been published & validated for use in ELISA & WB.

Physical form

Affinity purified immunoglobulin from pA(AB) 4-agarose column. IgG fraction in 10 mM sodium phosphate buffer (pH 7.4), 0.15 M NaCl, and 0.1% sodium azide.
Format: Purified

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service