Skip to Content
Merck
All Photos(1)

Documents

MAB4388

Sigma-Aldrich

Mouse Anti-Stella Antibody

clone 3H5.2, Chemicon®, from mouse

Synonym(s):

Anti-Anti-Pgc7, Anti-Anti-STELLA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H5.2, monoclonal

species reactivity

mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunocytochemistry: suitable

input

sample type: human embryonic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type: mouse embryonic stem cell(s)

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Dppa3(73708)

General description

Stella is specifically expressed by oocytes, pluripotent cells and primordial germ cells at the time of their emergence in gastrulating embryos, and is an important maternal factor in the cleavage stages of mouse embryos (Bortvin, 2004). The requirement for stella in germ cell specification remains controversial, however, most embryos from stella deficient mice failed to develop normally and rarely reach the blastocyst stage (Bortvin, 2004; Payer, 2003). The stella gene encodes a protein with an SAP-like domain and splicing factor motif-like structure, suggesting possible roles in chromosomal organization and RNA processing (Payer, 2003).

Specificity

Reacts with stella from human and mouse.

Application

Mouse Anti-Stella Antibody detects level of Stella & has been published & validated for use in ELISA & IC.
Research Category
Stem Cell Research
Research Sub Category
Pluripotent & Early Differentiation

Physical form

Format: Purified
Purified Immunoglobulin presented as a liquid in PBS with 0.1% sodium azide.

Storage and Stability

Maintain refrigerated at 2-8C in undiluted aliquots for up to 12 months.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katja Linher et al.
PloS one, 4(12), e8263-e8263 (2009-12-17)
We have previously demonstrated that stem cells isolated from fetal porcine skin have the potential to form oocyte-like cells (OLCs) in vitro. However, primordial germ cells (PGCs), which must also be specified during the stem cell differentiation to give rise
Pengxiang Wang et al.
Cell proliferation, 57(1), e13533-e13533 (2023-08-04)
Primordial germ cells (PGCs) are the germline precursors that give rise to oocytes and sperm, ensuring the continuation of life. While the PGC specification is extensively studied, it remains elusive how the PGC population is sustained and expanded after they
Xun Xu et al.
Nature materials (2024-08-13)
Naive pluripotent stem cells have the highest developmental potential but their in vivo existence in the blastocyst is transient. Here we report a blastocyst motif substrate for the in vitro reversion of mouse and human pluripotent stem cells to a
Maurizio Zuccotti et al.
BMC developmental biology, 8, 97-97 (2008-10-08)
The maternal contribution of transcripts and proteins supplied to the zygote is crucial for the progression from a gametic to an embryonic control of preimplantation development. Here we compared the transcriptional profiles of two types of mouse MII oocytes, one
Wenlin Li et al.
Methods in molecular biology (Clifton, N.J.), 1074, 31-37 (2013-08-27)
The derivation of mouse embryonic stem cells (mESCs) from the inner cell mass (ICM) of mouse preimplantation blastocysts has provided tremendous opportunities in exploring mammalian development and pluripotent stem cell biology (Martin, Proc Natl Acad Sci U S A 78(12):7634-7638

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service