Skip to Content
Merck
All Photos(1)

Key Documents

MAB3291

Sigma-Aldrich

Anti-HRAS Antibody

CHEMICON®, mouse monoclonal, 7D7.2

Synonym(s):

Anti-C-BAS/HAS, Anti-C-H-RAS, Anti-C-HA-RAS1, Anti-CTLO, Anti-H-RASIDX, Anti-HAMSV, Anti-HRAS1, Anti-RASH1, Anti-p21ras

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$738.00

$738.00


Estimated to ship on23 April 2025


Request a Bulk Order

Select a Size

Change View
100 μG
$738.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$738.00


Estimated to ship on23 April 2025


Request a Bulk Order

Product Name

Anti-H-Ras Antibody, clone 7D7.2, clone 7D7.2, Chemicon®, from mouse

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7D7.2, monoclonal

species reactivity

rat, mouse, human

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2b

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HRAS(3265)

Specificity

Recognizes H-Ras (the oncogene of Harvey murine sarcoma virus). Approximate molecular weight of 21 kDa.

Immunogen

Full-length recombinant human H-Ras - GST fusion protein

Application

Research Category
Signaling
Research Sub Category
MAP Kinases
This Anti-H-Ras Antibody, clone 7D7.2 is validated for use in ELISA, WB for the detection of H-Ras.
Western blot

ELISA

Optimal working dilutions must be determined by end user.

Physical form

0.02M phosphate buffer, 0.25M NaCl, pH 7.6, 0.1% sodium azide
Format: Purified

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 6 months.

WARNING: The monoclonal reagent solution contains 0.1% sodium azide as a preservative. Due to potential hazards arising from the build up of this material in pipes, spent reagent should be disposed of with liberal volumes of water.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ho-June Lee et al.
Molecular cancer therapeutics, 16(4), 694-704 (2017-02-01)
Cancer cell line profiling to identify previously unrecognized kinase dependencies revealed a novel nonmutational dependency on the DNA damage response checkpoint kinase Chk1. Although Chk1 is a promising therapeutic target in p53-deficient cancers, we found that Ras-MEK signaling engages Chk1
Wouter W Kallemeijn et al.
Nature protocols, 16(11), 5083-5122 (2021-10-29)
Protein lipidation is one of the most widespread post-translational modifications (PTMs) found in nature, regulating protein function, structure and subcellular localization. Lipid transferases and their substrate proteins are also attracting increasing interest as drug targets because of their dysregulation in
Candida Zuchegna et al.
Redox report : communications in free radical research, 27(1), 150-157 (2022-07-14)
Although the protooncogenes small GTPases Ras are redox-sensitive proteins, how they are regulated by redox signaling in the central nervous system (CNS) is still poorly understood. Alteration in redox-sensitive targets by redox signaling may have myriad effects on Ras stability
Atsushi Hoshino et al.
Nature communications, 4, 2308-2308 (2013-08-07)
Cumulative evidence indicates that mitochondrial dysfunction has a role in heart failure progression, but whether mitochondrial quality control mechanisms are involved in the development of cardiac dysfunction remains unclear. Here we show that cytosolic p53 impairs autophagic degradation of damaged

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service