Skip to Content
Merck
All Photos(1)

Documents

MAB13429

Sigma-Aldrich

Anti-TIMP-1 Antibody, clone 102D1

clone 102D1, Chemicon®, from mouse

Synonym(s):

Tissue Inhibitor of Metalloproteinase-1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

102D1, monoclonal

species reactivity

human

manufacturer/tradename

Chemicon®

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TIMP1(7076)

Specificity

It recognizes a glycoprotein of 28.5 kDa which is identified as tissue inhibitor of metalloproteinases-1 (TIMP-1). TIMP-1 and TIMP-2 have similar properties, specifically in inhibiting enzymes of matrix metalloproteinase family, and are thought to be of great importance in the maintenance of connective tissue integrity. TIMP-1 forms a complex of 1:1 stoichiometry with activated interstitial collagenase, activated stromelysin, active form of 72 kDa type IV collagenase (also known as MMP-2 or gelatinase A), and latent and active forms 92 kDa Type IV collagenase (also known as MMP-9 or gelatinase B). TIMPs inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells.

Cellular Localization:

Cytoplasmic

Immunogen

Recombinant human TIMP-1.

Application

Anti-TIMP-1 Antibody, clone 102D1 is an antibody against TIMP-1 for use in WB, IH(P).
Research Category
Cell Structure
Research Sub Category
MMPs & TIMPs
Western Blotting: 1-2 μg/mL for 2 hours at room temperature

Immunohistochemistry: Formalin-fixed, paraffin-embedded tissues: 1-2 μg/mL for 30 min. at room temperature.

Optimal working dilutions must be determined by end user.

Physical form

Format: Purified
Purified from ascites fluid by Protein G chromatography. Liquid in 10 mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 12 months after date of receipt.

Analysis Note

Control
POSITIVE CONTROL: Cytotrophoblastic columns, the endothelial and fibroblastic stromal cells of villi, and the large decidualized cells of decidual membrane in placenta1.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takahiko Naka et al.
Cancer, 112(1), 104-110 (2007-10-24)
Hepatocyte growth factor (HGF) is a multipotent cytokine that is mediated by its receptor, c-MET. HGF/c-MET contributes to tumor progression in many human malignancies; however, HGF/c-MET is inversely correlated with aggressive biologic behavior in other cancers. Conversely, to the authors'
David Moreno-Ajona et al.
BMC cardiovascular disorders, 20(1), 93-93 (2020-02-27)
Major adverse cardiovascular events are the main cause of morbidity and mortality over the long term in patients undergoing carotid endarterectomy. There are few reports assessing the prognostic value of markers of inflammation in relation to the risk of cardiovascular
Yuko Sugiyama et al.
Gastroenterology, 128(2), 480-486 (2005-02-03)
The surrounding cancer stroma is increasingly recognized as playing an important role in cancer proliferation, invasion, and metastasis. Here, we analyzed patterns of gene expression in colon cancer cells, surrounding stroma, and normal mucosa and normal stroma using laser capture
Alessia Frati et al.
Molecular endocrinology (Baltimore, Md.), 29(1), 53-67 (2014-11-22)
The initiation and progression of heart failure is linked to adverse cardiac remodeling of the extracellular matrix (ECM) during disease mainly through the deregulation of myocardial metalloproteinases (MMPs). Relaxin (RLX), a peptide hormone acting as a physiological cardiac effector, is
Pannatas Seanpong et al.
Japanese journal of infectious diseases, 68(3), 221-229 (2015-02-13)
Disease severities following dengue virus (DV) infection are the result of increased vascular permeability leading to hypovolemic shock. Matrix metalloproteinases (MMPs) are believed to play a key role in promoting such severities. A previous study reported that supernatants of DV-infected

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service