Skip to Content
Merck
All Photos(1)

Key Documents

FCMAB196P

Sigma-Aldrich

Milli-Mark® Anti-CD42b-PE Antibody, clone AN51

clone AN51, Milli-Mark®, from mouse

Synonym(s):

Platelet glycoprotein Ib alpha chain, Glycoprotein Ibalpha, GP-Ib alpha, GPIb-alpha, GPIbA, Antigen CD42b-alpha

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.44

biological source

mouse

Quality Level

conjugate

PE

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

AN51, monoclonal

species reactivity

human

manufacturer/tradename

Milli-Mark®

technique(s)

flow cytometry: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GP1BA(2811)

General description

CD42b is a 145 kDa protein, which together with CD42c forma a 160 kDa heterodimer composed of an alpha-chain and a beta-chain, respectively. The subunits are linked together by a disulphide bond. CD42b and CD42c forms a non-covalent complex together with CD42a and CD42d in the platelet plasma membrane. The CD42 complex serves as a receptor for von Willebrand factor and thrombin and mediates adhesion of platelets to subendothelial matrices (exposed upon damage to the endothelium) at high shear rates. The binding sites for von Willebrand factor and thrombin lies on CD42b (de Haas, 2002). CD42 is expressed on megakarytocytes and platelets (McMichael, 1981).

Specificity

Antibody recognizes Human CD42b antigen.

Immunogen

Mixture of human platelets and lymphocytes.

Application

Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology
This Milli-Mark Anti-CD42b-PE Antibody, clone AN51 is validated for use in FC for the detection of CD42b.

Quality

Evaluated by flow cytometry using human lymphocytes

Target description

145 kDa (de Haas, 2002)

Physical form

Protein A purified
Purified Mouse monoclonal IgG2a conjugated to PE in 0.05M Tris-HCl buffer with 1% BSA, Sodium Azide at 0.097% at pH 7.2.

Storage and Stability

Maintain refrigerated at 2-8°C protected from light in undiluted aliquots for up to 6 months from date of receipt.

Analysis Note

Control
Human Lymphocytes

Legal Information

MILLI-MARK is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service