Skip to Content
Merck
All Photos(1)

Key Documents

DX1045

Supelco

Diethylamine

Synonym(s):

Diethylamine, Diethamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2)2NH
CAS Number:
MDL number:
UNSPSC Code:
12352116
EC Index Number:
203-716-3

vapor pressure

253 hPa ( 20 °C)

Quality Level

Assay

≥98%

form

liquid

impurities

≤1000 ppm Water (H2O)

color

APHA: ≤30

pH

13 (20 °C, 100 g/L in H2O)

bp

56 °C/1013 mbar

mp

-50 °C

transition temp

flash point -23 °C

density

0.71 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

InChI

1S/C4H11N/c1-3-5-4-2/h5H,3-4H2,1-2H3

InChI key

HPNMFZURTQLUMO-UHFFFAOYSA-N

Analysis Note

Assay: 98% min
Color (APHA): 30 max
Form: Clear liquid
Water (H2O): 1000ppm max

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-14.8 °F - closed cup

Flash Point(C)

-26 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service