Skip to Content
Merck
All Photos(5)

Key Documents

ABE48

Sigma-Aldrich

Anti-NELF-E Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Negative elongation factor E, NELF-E, RNA-binding protein RD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human, mouse

species reactivity (predicted by homology)

canine (based on 100% sequence homology), bovine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... NELFE(7936)

General description

NELF-E (Negative elongation factor E) is a main component of NELF human transcription factor complex which associates with DRB sensitivity-inducing factor to stifle transcriptional elongation by RNA polymerase II. NELF-E is an RNA-binding protein that includes arginine-aspartic acid dipeptide repeats. NELF-E RNA-binding activity is essential for NELF to function.

Immunogen

KLH-conjugated linear peptide corresponding to human NELF-E.

Application

Anti-NELF-E Antibody is a Rabbit Polyclonal Antibody for detection of NELF-E also known as Negative elongation factor E, NELF-E, RNA-binding protein RD & has been validated in ICC & WB.
Immunocytochemistry Analysis: 1:500 dilution from a representative lot detected NELF-E in NIH/3T3 and HeLa cells.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Quality

Evaluated by Western Blot in L6 cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected NELF-E on 10 µg of L6 cell lysate.

Target description

~ 43 kDa observed. Uncharacterized bands may be due to post-translational modification.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
L6 cell lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Integrator complex regulates NELF-mediated RNA polymerase II pause/release and processivity at coding genes.
Stadelmayer, B; Micas, G; Gamot, A; Martin, P; Malirat, N; Koval, S; Raffel, R; Sobhian et al.
Nature Communications null
Seychelle M Vos et al.
eLife, 5 (2016-06-11)
Transcription regulation in metazoans often involves promoter-proximal pausing of RNA polymerase (Pol) II, which requires the 4-subunit negative elongation factor (NELF). Here we discern the functional architecture of human NELF through X-ray crystallography, protein crosslinking, biochemical assays, and RNA crosslinking

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service