Skip to Content
Merck
All Photos(2)

Key Documents

ABC827

Sigma-Aldrich

Anti-IFITM1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Interferon-induced transmembrane protein 1, Dispanin subfamily A member 2a, DSPA2a, Interferon-induced protein 17, Interferon-inducible protein 9-27, Leu-13 antigen, CD225

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$661.00

$661.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$661.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$661.00


Estimated to ship on24 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse, human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IFITM1(8519)

General description

Interferon-induced transmembrane protein 1, also known as Dispanin subfamily A member 2a (DSPA2a) or Interferon-induced protein 17 or Interferon-inducible protein 9-27 or Leu-13 antigen or CD225, and encoded by the gene name IFITM1 or CD225 is a member of the IFN-inducible transmembrane protein family. It is an essential mediator of interferon-γ-induced antiproliferation and plays a role in the control of cell growth and migration. IFITM1 is upregulated in several tumor types and may be useful as a tumor biomarker. Both mouse IFITM1 and IFITM3 are expressed on the cell surfaces of primordial germ cells in a developmentally-regulated manner. IFITM1 activity is required for primordial germ cell transit, and IFITM1 acts as a repulsive molecule by repelling non-IFITM1-expressing primordial germ cells from the mesoderm into the endoderm.

Specificity

This antibody will cross-react with IFITM2 and IFITM3.

Immunogen

Epitope: center region
KLH-conjugated synthetic peptide corresponding to the center region of human IFITM1.

Application

Immunocytochemistry Analysis: 20 μg/mL from a representative lot detected IFITM1 in Jurkat cells.

Immunofluorescence Analysis: 20 μg/mL from a representative lot detected IFITM1 in Jurkat cells.
Research Category
Apoptosis & Cancer
Research Sub Category
Apoptosis - Additional
This Anti-IFITM1 Antibody is validated for use in western blotting, ICC & immunofluorescence for the detection of IFITM1.

Quality

Evaluated by Western Blotting in NIH/3T3 cell lysate.

Western Blotting Analysis: 2 μg/mL of this antibody detected IFITM1 in NIH/3T3 cell lysate.

Target description

~14 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Antigen Affinity Purified
Purified rabbit polyclonal in buffer containing PBS with up to 0.1% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service