Skip to Content
Merck
All Photos(1)

Documents

AB15452

Sigma-Aldrich

Anti-MAP2 Antibody

Chemicon®, from chicken

Synonym(s):

Microtubule associated protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

chicken

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

mouse

manufacturer/tradename

Chemicon®

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Map2(17756)

Specificity

MAP2 [Microtubule associated protein 2]
Other species have not yet been tested. It is expected that the antibody will also react with human due to immunogen sequence similarity.

Immunogen

Synthetic peptides corresponding to different regions of the human/mouse MAP2 protein.

Application

Anti-MAP2 Antibody detects level of MAP2 & has been published & validated for use in IC, IH.
Immunohistochemistry: 1:1000-1:2,000 using 2% paraformaldehyde fixed tissue.

Immunocytochemistry: 1:1000-1:2,000 using 2% paraformaldehyde fixed cells.

Optimal working dilutions must be determined by end user.
Research Category
Neuroscience
Research Sub Category
Neuronal & Glial Markers

Neurofilament & Neuron Metabolism

Physical form

Affinity purified chicken immunoglobulin. Two different antibodies were combined to make this product. Liquid in PBS, pH 7.2 with 0.02% sodium azide.
ImmunoAffinity Purified

Storage and Stability

Maintain at 2-8°C in undiluted aliquots for up to 6 months after date of receipt.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yun Yang et al.
Nature communications, 13(1), 159-159 (2022-01-12)
Abnormalities in brain glucose metabolism and accumulation of abnormal protein deposits called plaques and tangles are neuropathological hallmarks of Alzheimer's disease (AD), but their relationship to disease pathogenesis and to each other remains unclear. Here we show that succinylation, a
Andrew R Snavely et al.
Disease models & mechanisms, 15(12) (2022-11-19)
The proteosome inhibitor bortezomib has revolutionized the treatment of multiple hematologic malignancies, but in many cases, its efficacy is limited by a dose-dependent peripheral neuropathy. We show that human induced pluripotent stem cell (hiPSC)-derived motor neurons and sensory neurons provide
Hae-Jin Kweon et al.
Cell reports, 32(6), 108025-108025 (2020-08-14)
The α7 nicotinic acetylcholine receptor participates in diverse aspects of brain physiology and disease. Neurons tightly control α7 assembly, which relies upon NACHO, an endoplasmic reticulum (ER)-localized integral membrane protein. By constructing α7 chimeras and mutants, we find that NACHO
Madhurima Dhara et al.
Nature communications, 11(1), 2799-2799 (2020-06-05)
Small molecule polyamines are abundant in all life forms and participate in diverse aspects of cell growth and differentiation. Spermidine/spermine acetyltransferase (SAT1) is the rate-limiting enzyme in polyamine catabolism and a primary genetic risk factor for suicidality. Here, using genome-wide
Maria Vedunova et al.
Frontiers in cellular neuroscience, 7, 149-149 (2013-09-26)
The extracellular matrix (ECM) plays an important role in use-dependent synaptic plasticity. Hyaluronic acid (HA) is the backbone of the neural ECM, which has been shown to modulate α-amino-3-hydroxyl-5-methyl-4-isoxazolepropionate (AMPA) receptor mobility, paired-pulse depression, L-type voltage-dependent Ca(2+) channel (L-VDCC) activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service