Skip to Content
Merck
All Photos(2)

Key Documents

16-224

Sigma-Aldrich

Anti-Myc Tag Antibody, clone 4A6, Alexa Fluor 488 conjugate

clone 4A6, Upstate®, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

mouse

Quality Level

conjugate

ALEXA FLUOR 488

antibody product type

primary antibodies

clone

4A6, monoclonal

species reactivity

human

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
western blot: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Specificity

Other species cross-reactivity not tested.
Recognizes and is specific for recombinant proteins containing the Myc epitope tag (EQKLISEEDL) in a variety of sequence contexts. Also recognizes human Myc.

Immunogen

KLH-conjugated, synthetic peptide corresponding to amino acids 410-420 (MEQKLISEEDL) of human Myc

Application

Anti-Myc Tag Antibody, clone 4A6, Alexa Fluor 488 conjugate is an antibody against Myc Tag for use in IF, WB & IC.
Research Category
Epitope Tags & General Use
Research Sub Category
Epitope Tags

Quality

Routinely evaluated by immunoblot and immunocytochemistry

Target description

Varies

Legal Information

ALEXA FLUOR is a trademark of Life Technologies
UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Alexa Fluor is a registered trademark of Molecular Probes, Inc.

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hirotomo Nakahara et al.
Cancer immunology research, 1(4), 223-228 (2014-01-17)
For antigen recognition, lampreys use leucine-rich repeats (LRR) instead of immunoglobulin V-(D)-J domains to generate variable lymphocyte receptors (VLR) of three types, VLRA, VLRB, and VLRC. VLRB-bearing lymphocytes respond to immunization with proliferation and differentiation into plasmacytes that secrete multivalent
Alexis J Lomakin et al.
Nature cell biology, 17(11), 1435-1445 (2015-09-29)
Symmetry-breaking polarization enables functional plasticity of cells and tissues and is yet not well understood. Here we show that epithelial cells, hard-wired to maintain a static morphology and to preserve tissue organization, can spontaneously switch to a migratory polarized phenotype
André C Velásquez et al.
Plant methods, 13, 29-29 (2017-04-22)
The ability to target and manipulate protein-based cellular processes would accelerate plant research; yet, the technology to specifically and selectively target plant-expressed proteins is still in its infancy. Leucine-rich repeats (LRRs) are ubiquitously present protein domains involved in mediating protein-protein
Lars Andresen et al.
Journal of immunology (Baltimore, Md. : 1950), 188(4), 1847-1855 (2012-01-10)
NKG2D ligand surface expression is important for immune recognition of stressed and neotransformed cells. In this study, we show that surface expression of MICA/B and other NKG2D ligands is dependent on N-linked glycosylation. The inhibitor of glycolysis and N-linked glycosylation
Catarina A Marques et al.
Nucleic acids research, 44(10), 4763-4784 (2016-03-10)
Initiation of DNA replication depends upon recognition of genomic sites, termed origins, by AAA+ ATPases. In prokaryotes a single factor binds each origin, whereas in eukaryotes this role is played by a six-protein origin recognition complex (ORC). Why eukaryotes evolved

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service