Skip to Content
Merck
All Photos(1)

Documents

14-452-D

Sigma-Aldrich

PDK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.27

description

protein target PDK1

Quality Level

Assay

≥80%

specific activity

5.02 pmol/min-μg

concentration

0.5 mg/mL

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... PDPK1(5170)

General description

FUNCTION: SwissProt: O15530 # Phosphorylates and activates not only PKB/AKT, but also PKA, PKC-zeta, RPS6KA1 and RPS6KB1. May play a general role in signaling processes and in development (By similarity). Isoform 3 is catalytically inactive.
SIZE: 556 amino acids; 63152 Da
SUBCELLULAR LOCATION: Cytoplasm. Membrane; Peripheral membrane protein. Note=Membrane-associated after cell stimulation leading to its translocation. Tyrosine phosphorylation seems to occur only at the plasma membrane.
TISSUE SPECIFICITY: Appears to be expressed ubiquitously.
PTM: Phosphorylated on tyrosine and serine/threonine. Phosphorylation on Ser-241 in the activation loop is required for full activity. PDK1 itself can autophosphorylate Ser-241, leading to its own activation.
SIMILARITY: SwissProt: O15530 ## Belongs to the protein kinase superfamily. AGC Ser/Thr protein kinase family. PDK1 subfamily. & Contains 1 PH domain. & Contains 1 protein kinase domain.

Quality

Evaluated by Kinase assay: Enzyme reaction is conducted in a buffer containing 50 mM HEPES (pH7.5), 10 mM MgCl2, 1 mM EGTA, 200 EM ATP, 0.01% Brij-35 and 2 EM substrate (SER/THR 6, Invitrogen) at room temperature for 1 hour.

Physical properties

89 kDa

Physical form

40 mM Tris-HCl, pH 8.0, 110 mM NaCl, 2.2 mM KCl, 20% glycerol, and 3 mM DTT

Storage and Stability

Stable for > 6 month at –80°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service