Skip to Content
Merck
All Photos(1)

Key Documents

07-892

Sigma-Aldrich

Anti-phospho-Pyk2/CAKβ/FAK2 (Tyr402) Antibody

Upstate®, from rabbit

Synonym(s):

PTK2B, Proline-rich tyrosine kinase 2, Focal adhesion kinase 2, FADK2, Cell adhesion kinase beta, RAFTK, Calcium-dependent tyrosine kinase, CADTK

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$781.00

$781.00


Estimated to ship on24 April 2025


Request a Bulk Order

Select a Size

Change View
100 μL
$781.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

$781.00


Estimated to ship on24 April 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, rat

manufacturer/tradename

Upstate®

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

dry ice

target post-translational modification

phosphorylation (pTyr402)

Gene Information

human ... PTK2B(2185)

Specificity

Pyk2 phosphorylated on tyrosine residue 402

Immunogen

Peptide corresponding to amino acid region encompassing the human and rat phospho-Pyk2/CAKβ/FAK2 (Tyr402).

Application

Immunoblot Analysis: 1:1000 dilutions of this lot detected phospho-Pyk2 (Tyr402) in RIPA lysate of Pyk2 transfected CEF cells
This Anti-phospho-Pyk2/CAKβ/FAK2 (Tyr402) Antibody is validated for use in WB, IC for the detection of phospho-Pyk2/CAKβ/FAK2 (Tyr402).

Quality

Routinely evaluated by immunoblot.

Target description

115 kDa

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M D Schaller et al.
The Journal of biological chemistry, 272(40), 25319-25325 (1997-10-06)
pp125(FAK) and CAKbeta/Pyk2/CadTK/RAFTK are related protein-tyrosine kinases. It is therefore of interest whether CAKbeta shares some of the properties of pp125(FAK). Using recombinant glutathione S-transferase fusion proteins, we show that the C-terminal domains of both proteins bind paxillin in vitro.
Monika Raab et al.
Nature communications, 8, 16001-16001 (2017-07-13)
Lymphocyte function-associated antigen 1 (LFA-1) affinity and avidity changes have been assumed to mediate adhesion to intercellular adhesion molecule-1 for T-cell conjugation to dendritic cells (DC). Although the T-cell receptor (TCR) and LFA-1 can generate intracellular signals, the immune cell

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service