Skip to Content
Merck
All Photos(1)

Key Documents

MAT246517598

Matrix Scientific

4-Chloro-6-(3-piperidinylmethoxy)pyrimidinehydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$379.29

Vendor SKU: 050875-500MG

$379.29


Ships in 7 days - from Aldrich Partner.


Select a Size

Change View
500 MG
$379.29

About This Item

CAS Number:
Molecular Weight:
264.15
MDL number:

Vendor SKU: 050875-500MG

$379.29


Ships in 7 days - from Aldrich Partner.

Iupac Name

4-chloro-6-(3-piperidinylmethoxy)pyrimidine hydrochloride

Inchi Code

1S/C10H14ClN3O.ClH/c11-9-4-10(14-7-13-9)15-6-8-2-1-3-12-5-8;/h4,7-8,12H,1-3,5-6H2;1H

InChI key

ISUWNGVCLFKSBU-UHFFFAOYSA-N

Country of Origin

US

Product Link

https://www.matrixscientific.com/050875.html

Pictograms

MSDS

https://www.matrixscientific.com/050875.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service