MAT161065775
Matrix Scientific
3-[(3-Hydroxypropyl)amino]propanenitrile
Select a Size
Vendor SKU: 026771-500MG
$486.93
Select a Size
About This Item
Vendor SKU: 026771-500MG
$486.93
bp
110-115
Iupac Name
3-[(3-hydroxypropyl)amino]propanenitrile
Inchi Code
1S/C6H12N2O/c7-3-1-4-8-5-2-6-9/h8-9H,1-2,4-6H2
InChI key
YTMXVUVIZPTHBO-UHFFFAOYSA-N
Country of Origin
Product Link
Boiling point
MSDS
https://www.matrixscientific.com/026771.html
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service