Skip to Content
Merck
All Photos(1)

Key Documents

KEY423214583

Key Organics / BIONET

methyl 6-oxo-1-{[4-(trifluoromethyl)phenyl]methyl}-1,6-dihydropyridine-3-carboxylate

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$363.94
1 G
$527.81

Vendor SKU: 6E-368S-500MG

$363.94


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
$363.94
1 G
$527.81

About This Item

CAS Number:
Molecular Weight:
311.26
MDL number:

Vendor SKU: 6E-368S-500MG

$363.94


Ships in 21 days - from Aldrich Partner.

mp

103 - 105

Iupac Name

methyl 6-oxo-1-(4-(trifluoromethyl)benzyl)-1,6-dihydropyridine-3-carboxylate

Inchi Code

1S/C15H12F3NO3/c1-22-14(21)11-4-7-13(20)19(9-11)8-10-2-5-12(6-3-10)15(16,17)18/h2-7,9H,8H2,1H3

InChI key

WQGGROBXGHKZSO-UHFFFAOYSA-N

Purity

97%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/6E-368S

Melting Point

103 - 105

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/6E-368S


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service