Skip to Content
Merck
All Photos(1)

Key Documents

JWPH3249D4AA

J & W Pharmlab LLC

4,6-Difluoro-2,3-dihydro-1H-isoindole

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
$501.94
100 MG
$717.56
250 MG
$1,148.81
500 MG
$1,580.06
1 G
$2,226.94
5 G
$8,695.69

Vendor SKU: 48R0845-50MG

$501.94


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
$501.94
100 MG
$717.56
250 MG
$1,148.81
500 MG
$1,580.06
1 G
$2,226.94
5 G
$8,695.69

About This Item

CAS Number:
Molecular Weight:
155.15
MDL number:

Vendor SKU: 48R0845-50MG

$501.94


Ships in 14 days - from Aldrich Partner.

Iupac Name

4,6-difluoroisoindoline

Inchi Code

1S/C8H7F2N/c9-6-1-5-3-11-4-7(5)8(10)2-6/h1-2,11H,3-4H2

InChI key

FWBHIHDAOIYJOE-UHFFFAOYSA-N

Purity

98%

Country of Origin

US

Physical Form

Light Pink Solid

Product Link

https://www.jwpharmlab.com/home/product/48R0845

MSDS

https://www.jwpharmlab.com/home/product/48R0845


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service