Skip to Content
Merck

ENAH97BD0635

Enamine

1-methyl-4-(propan-2-yl)cyclohex-1-ene

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
138.25
MDL number:

Pricing and availability is not currently available.

Iupac Name

4-isopropyl-1-methyl-1-cyclohexene

Inchi Code

1S/C10H18/c1-8(2)10-6-4-9(3)5-7-10/h4,8,10H,5-7H2,1-3H3

InChI key

FAMJUFMHYAFYNU-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-7182893

MSDS

https://www.enaminestore.com/catalog/EN300-7182893


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service