Skip to Content
Merck
All Photos(1)

Key Documents

ENAH942F3082

Enamine

3-methoxy-2-(methoxymethyl)propane-1-sulfonyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
216.69
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

3-methoxy-2-(methoxymethyl)propane-1-sulfonyl chloride

Inchi Code

1S/C6H13ClO4S/c1-10-3-6(4-11-2)5-12(7,8)9/h6H,3-5H2,1-2H3

InChI key

CNESYTMHJZLOOD-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-245322

MSDS

https://www.enaminestore.com/catalog/EN300-245322


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service