ENAH03BB375A
Enamine
3,5-dibromoimidazo[1,2-a]pyrazine
Select a Size
Vendor SKU: 10T-0605-500MG
EUR 107.80
Select a Size
About This Item
Vendor SKU: 10T-0605-500MG
EUR 107.80
mp
150-151
Iupac Name
3,5-dibromoimidazo[1,2-a]pyrazine
Inchi Code
1S/C6H3Br2N3/c7-4-1-9-3-6-10-2-5(8)11(4)6/h1-3H
InChI key
UVTHYCYGHSMBEV-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Melting Point
MSDS
https://www.enaminestore.com/catalog/EN300-749190
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service