Skip to Content
Merck
All Photos(1)

Key Documents

ENA369382725

Enamine

3,5-bis(propan-2-yl)-4H-1,2,4-triazole

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
153.23
MDL number:

mp

154-155

Iupac Name

3,5-diisopropyl-4H-1,2,4-triazole

Inchi Code

1S/C8H15N3/c1-5(2)7-9-8(6(3)4)11-10-7/h5-6H,1-4H3,(H,9,10,11)

InChI key

GBXKHQMLNCTDEG-UHFFFAOYSA-N

storage temp.

RT

Purity

93

Country of Origin

UA

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-35659

Melting Point

154-155

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-35659


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service